Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibromopentane 98.0+%, TCI America™
SDP

Catalog No. D14105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D14105G Supplier TCI America Supplier No. D14105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19398-53-9
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.94
MDL Number MFCD00039176
InChI Key CITMYAPULDSOHG-UHFFFAOYNA-N
PubChem CID 137228
IUPAC Name 1,2-dibromopentane
SMILES CCCC(Br)CBr

Specifications

Color Yellow
Boiling Point 40°C
Quantity 5 g
UN Number 1993
Formula Weight 229.94
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dibromopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.