accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHBrF

CAS RN 194019-11-9

IUPAC Name: 2-(3-bromo-4-fluorophenyl)acetic acid
Synonyms: (3-bromo-4-fluorophenyl)acetic acid
Molecular Weight (g/mol): 233.04
Molecular Formula: C8H6BrFO2
InChi Key: XXFGIJYSXNXNAU-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(Br)=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)