accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHOCH3CH3HO

CAS RN 19408-84-5

IUPAC Name: N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnonanamide
Synonyms: dihydrocapsaicin 6,7-dihydrocapsaicin
Molecular Weight (g/mol): 307.43
Molecular Formula: C18H29NO3
InChi Key: XJQPQKLURWNAAH-UHFFFAOYSA-N
SMILES: COC1=CC(CNC(=O)CCCCCCC(C)C)=CC=C1O
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)