CAS RN 19423-78-0
CAS RN 19423-78-0
IUPAC Name:
neodymium(3+) trichloride
Synonyms:
neodymium(III) chloride
Molecular Weight (g/mol):
250.59
Molecular Formula:
Cl3Nd
InChi Key:
ATINCSYRHURBSP-UHFFFAOYSA-K
SMILES:
[Cl-].[Cl-].[Cl-].[Nd+3]
1
–
2
of
2
results
1
Neodymium(III) chloride hydrate, REacton™, 99.9% (REO)
CAS: 19423-78-0 MDL Number: MFCD00011132
2
Neodymium(III) chloride hydrate, REacton™, 99.99% (REO)
CAS: 19423-78-0 MDL Number: MFCD00011132