accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OClClClGd

CAS RN 19423-81-5

IUPAC Name: gadolinium(3+) hydrate trichloride
Synonyms: gadolinium(3+) hydrate trichloride
Molecular Weight (g/mol): 281.62
Molecular Formula: Cl3GdH2O
InChi Key: SXVCBXHAHBCOMV-UHFFFAOYSA-K
SMILES: O.[Cl-].[Cl-].[Cl-].[Gd+3]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results