accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19423-86-0

IUPAC Name: thulium(3+) trichloride
Synonyms: thulium(III) chloride
Molecular Weight (g/mol): 275.28
Molecular Formula: Cl3Tm
InChi Key: ILOTUXNTERMOJL-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Tm+3]
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)