accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOHOO

CAS RN 19506-87-7

IUPAC Name: 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
Synonyms: 2-(1,3-dioxoisoindol-2-yl)propanoic acid
Molecular Weight (g/mol): 219.2
Molecular Formula: C11H9NO4
InChi Key: OZWUITKBAWTEAQ-UHFFFAOYNA-N
SMILES: CC(N1C(=O)C2=CC=CC=C2C1=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results