Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthaloyl-DL-alanine 99.0+%, TCI America™
SDP

Catalog No. P069525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P069525G 25 g
P069510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P069525G Supplier TCI America Supplier No. P069525G

Chemical Identifiers

CAS 19506-87-7
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.20
MDL Number MFCD00023061,MFCD00190990
InChI Key OZWUITKBAWTEAQ-UHFFFAOYNA-N
Synonym Phthalyl-DL-alanine
PubChem CID 86884
IUPAC Name 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
SMILES CC(N1C(=O)C2=CC=CC=C2C1=O)C(O)=O

Specifications

Melting Point 163°C
Color White
Quantity 25 g
Formula Weight 219.20
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phthaloyl-DL-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.