accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19522-67-9

IUPAC Name: (2-aminoethyl)(propan-2-yl)amine
Synonyms: (2-aminoethyl)(isopropyl)amine
Molecular Weight (g/mol): 102.18
Molecular Formula: C5H14N2
InChi Key: KDRUIMNNZBMLJR-UHFFFAOYSA-N
SMILES: CC(C)NCCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results