accessibility menu, dialog, popup

UserName

Viewing profile as user:
KOOHN

CAS RN 19525-59-8

IUPAC Name: potassium 2-(phenylamino)acetate
Synonyms: potassium DL-α-phenylaminoacetate
Molecular Weight (g/mol): 189.26
Molecular Formula: C8H8KNO2
InChi Key: JINONCVJYBHUCP-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)CNC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results