accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1953-99-7

IUPAC Name: tetrachlorobenzene-1,2-dicarbonitrile
Synonyms: tetrachlorobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 265.9
Molecular Formula: C8Cl4N2
InChi Key: OQHXZZGZASQSOB-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(Cl)=C(C#N)C(C#N)=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results