Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorophthalonitrile 96.0+%, TCI America™
SDP

Catalog No. T30675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30675G 5 g
T306725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30675G Supplier TCI America Supplier No. T30675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1953-99-7
Molecular Formula C8Cl4N2
Molecular Weight (g/mol) 265.902
MDL Number MFCD00019740
InChI Key OQHXZZGZASQSOB-UHFFFAOYSA-N
Synonym Perchlorophthalonitrile
PubChem CID 16057
IUPAC Name 3,4,5,6-tetrachlorobenzene-1,2-dicarbonitrile
SMILES C(#N)C1=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C#N

Specifications

Melting Point 251°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3077
Formula Weight 265.90
Percent Purity ≥96.0% (GC,N)
Chemical Name or Material Tetrachlorophthalonitrile
EINECSNumber (3)-1783
RTECSNumber TI8225000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.