accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHCOOCHOOCHOOHCOZrCF3CF3CF3CF3CF3CF3CF3CF3

CAS RN 19530-02-0

IUPAC Name: zirconium(4+) tetrakis(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Synonyms: zirconium(4+) tetrakis(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Molecular Weight (g/mol): 919.43
Molecular Formula: C20H4F24O8Zr
InChi Key: JALYWUQUXGQVGB-UHFFFAOYSA-N
SMILES: [Zr+4].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results