Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) hexafluoro-2,4-pentanedionate, Thermo Scientific™

Catalog No. AA3954214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3954214 25 g
AA3954206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3954214 Supplier Thermo Scientific Chemicals Supplier No. 3954214

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19530-02-0
Molecular Formula C20H4F24O8Zr
Molecular Weight (g/mol) 919.43
MDL Number MFCD00079660
InChI Key JALYWUQUXGQVGB-UHFFFAOYSA-N
Synonym zirconium iv hexafluoroacetylacetonate, zirconium hexafluoropentanedionate, zirconium hexafluoroacetylacetonate, zirconium hexafluoroacetonylacetonate, zirconium iv hexafluoro-2,4-pentanedionate, zirconium iv hexafluoroacetylacetonate, ?zr cf3cochcocf3 4, 3z-1,1,1,5,5,5-hexafluoro-4-tris 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy zirconio oxy pent-3-en-2-one
PubChem CID 132274318
IUPAC Name (Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate;zirconium(4+)
SMILES [Zr+4].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F

Specifications

Boiling Point 225°C
Melting Point 40°C to 43°C
Color White
Physical Form Crystalline
Quantity 25 g
Formula Weight 919.47
Odor Odorless
Chemical Name or Material Zirconium(IV) hexafluoro-2,4-pentanedionate
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.