CAS RN 1955550-51-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ FMK 9a
Potent ATG4B/autophagin-1 inhibitor; also inhibits calpain and cathepsin B