accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1956-30-5

IUPAC Name: [3-(acetyloxy)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl](iodo)methyl acetate
Synonyms: [3-(acetyloxy)-5-(2,4-dioxo-3H-pyrimidin-1-yl)oxolan-2-yl](iodo)methyl acetate
Molecular Weight (g/mol): 438.17
Molecular Formula: C13H15IN2O7
InChi Key: AJHVMHGESCYBHX-UHFFFAOYNA-N
SMILES: CC(=O)OC(I)C1OC(CC1OC(C)=O)N1C=CC(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)