Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Diacetyl-5-iodo-2'-deoxyuridine, 99+%, Thermo Scientific™

Catalog No. AC226375000
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226375000 500 mg
AC226372500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226375000 Supplier Thermo Scientific Chemicals Supplier No. 226375000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1956-30-5
Molecular Formula C13H15IN2O7
Molecular Weight (g/mol) 438.174
MDL Number MFCD00063817
InChI Key LVDYIZDWXVFTQG-GMTAPVOTSA-N
Synonym 3',5'-di-o-acetyl-5-iodo-2'-deoxyuridine, 2'-deoxy-5-iodouridine 3',5'-diacetate, 3',5'-di-o-acetyl-2'-deoxy-5-iodouridine, 2r,3s,5r-3-acetoxy-5-5-iodo-2,4-dioxo-3,4-dihydropyrimidin-1 2h-yl tetrahydrofuran-2-yl methyl acetate, 2r,3s,5r-3-acetyloxy-5-5-iodo-2,4-dioxo-3h-pyrimidin-1-yl oxolan-2-yl methyl acetate, uridine,2'-deoxy-5-iodo-,3',5'-diacetate, 2r,3s,5r-3-acetyloxy-5-5-iodo-2,4-dioxopyrimidin-1-yl oxolan-2-yl methyl acetate
PubChem CID 21780160
IUPAC Name [(2R,3R,5R)-3-acetyloxy-5-(5-iodo-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(CC(O1)N2C=C(C(=O)NC2=O)I)OC(=O)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 500 mg
Formula Weight 438.16
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material 3', 5'-Diacetyl-5-iodo-2'-deoxyuridine, 99+%
Show More Show Less
EINECSNumber 217-799-9
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.