accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19576-08-0

IUPAC Name: 7a-methyl-2,3,5,6,7,7a-hexahydro-1H-indene-1,5-dione
Synonyms: 7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: FNYAZSZTENLTRT-UHFFFAOYNA-N
SMILES: CC12CCC(=O)C=C1CCC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results