accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1958-93-6

IUPAC Name: 2-(bromomethyl)-1-fluoro-3-nitrobenzene
Synonyms: 2-(bromomethyl)-1-fluoro-3-nitrobenzene
Molecular Weight (g/mol): 234.02
Molecular Formula: C7H5BrFNO2
InChi Key: KKSODTKRSQTJFZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(F)=C1CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results