Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-6-nitrobenzyl bromide, 98%, Thermo Scientific™

Catalog No. AC449150050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449150050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC449150050 Supplier Thermo Scientific Chemicals Supplier No. 449150050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 1958-93-6
Molecular Formula C7H5BrFNO2
Molecular Weight (g/mol) 234.02
MDL Number MFCD00042918
InChI Key KKSODTKRSQTJFZ-UHFFFAOYSA-N
PubChem CID 74779
IUPAC Name 2-(bromomethyl)-1-fluoro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(F)=C1CBr

Specifications

Melting Point 50°C to 54°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H3CH2Br(F)
Quantity 5 g
Synonym 2-fluoro-6-nitrobenzyl bromide, 2-bromomethyl-1-fluoro-3-nitrobenzene, benzene, 2-bromomethyl-1-fluoro-3-nitro, alpha-bromo-2-fluoro-6-nitrotoluene, acmc-20anuk, 2-fluoro-6-nitrobenzylbromide, 2-bromomethyl-3-fluoro-1-nitrobenzene, benzene,2-bromomethyl-1-fluoro-3-nitro
Solubility Information Solubility in water: insoluble
Formula Weight 234.02
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-nitrobenzyl bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 217-801-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.