accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19614-12-1

IUPAC Name: 1-bromo-2-(bromomethyl)-4-methoxybenzene
Synonyms: 1-bromo-2-(bromomethyl)-4-methoxybenzene
Molecular Weight (g/mol): 279.96
Molecular Formula: C8H8Br2O
InChi Key: MURVUTUZSUEIGI-UHFFFAOYSA-N
SMILES: COC1=CC(CBr)=C(Br)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)