Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methoxybenzyl bromide, 97%

Catalog No. AC439790250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC439790250 Supplier Thermo Scientific Chemicals Supplier No. 439790250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 19614-12-1 | C8H8Br2O | 279.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19614-12-1
Molecular Formula C8H8Br2O
Molecular Weight (g/mol) 279.96
InChI Key MURVUTUZSUEIGI-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzyl bromide, 1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 2-bromo-5-methoxy-benzyl bromide, benzene, 1-bromo-2-bromomethyl-4-methoxy, 4-bromo-3-bromomethyl-1-methoxybenzene, pubchem18176, acmc-20a5x2, 4-bromo3-bromomethylanisole
PubChem CID 3525951
IUPAC Name 1-bromo-2-(bromomethyl)-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)Br)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 91.0°C to 93.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 279.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methoxybenzyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.