accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19617-43-7

IUPAC Name: ethyl N-carbonylcarbamate
Synonyms: ethyl N-carbonylcarbamate
Molecular Weight (g/mol): 115.09
Molecular Formula: C4H5NO3
InChi Key: VMVZGGPZNHFGKS-UHFFFAOYSA-N
SMILES: CCOC(=O)N=C=O
Boiling Point 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)