Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl isocyanatoformate

Catalog No. 3082265G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-822-65G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30-822-65G Supplier Sigma Aldrich Supplier No. 3082265G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl isocyanatoformate

Specifications

CAS 19617-43-7
Density 1.115 g/mL (at 25°C (literature))
Boiling Point 25°C (10 mmHg)
Molecular Formula C4H5NO3
Linear Formula OCNCO2C2H5
Refractive Index n20/D 1.408 (literature)
MDL Number MFCD00011719
Quantity 250 mg
Synonym Ethoxycarbonyl isocyanate
Molecular Weight (g/mol) 115.09
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.