accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19634-89-0

IUPAC Name: 2,2'-dimethyl-1,1'-binaphthalene
Synonyms: 2,2'-dimethyl-1,1'-binaphthalene
Molecular Weight (g/mol): 282.39
Molecular Formula: C22H18
InChi Key: KDHFKMDVFWYSPT-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results