Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2,2'-Dimethyl-1,1'-binaphthyl 98.0+%, TCI America™
SDP

Catalog No. D28375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28375G 5 g
D28371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28375G Supplier TCI America Supplier No. D28375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19634-89-0
Molecular Formula C22H18
Molecular Weight (g/mol) 282.386
MDL Number MFCD00048075
InChI Key KDHFKMDVFWYSPT-UHFFFAOYSA-N
PubChem CID 141748
IUPAC Name 2-methyl-1-(2-methylnaphthalen-1-yl)naphthalene
SMILES CC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)C

Specifications

Melting Point 84°C
Color White
Quantity 5 g
Formula Weight 282.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-2,2′-Dimethyl-1,1′-binaphthyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.