accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNClCl

CAS RN 19646-07-2

IUPAC Name: 2,4-dichloro-5-methoxypyrimidine
Synonyms: 2,4-dichloro-5-methoxypyrimidine
Molecular Weight (g/mol): 179
Molecular Formula: C5H4Cl2N2O
InChi Key: ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
SMILES: COC1=CN=C(Cl)N=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)