accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNCl

CAS RN 19652-14-3

IUPAC Name: 3-(4-chlorophenyl)-1-phenyl-1H-pyrazol-5-amine
Synonyms: 5-(4-chlorophenyl)-2-phenylpyrazol-3-amine
Molecular Weight (g/mol): 269.73
Molecular Formula: C15H12ClN3
InChi Key: ZLGORAQNGFCUFU-UHFFFAOYSA-N
SMILES: NC1=CC(=NN1C1=CC=CC=C1)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results