Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole 98.0+%, TCI America™
SDP

Catalog No. A30335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30335G 5 g
A30331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30335G Supplier TCI America Supplier No. A30335G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19652-14-3
Molecular Formula C15H12ClN3
Molecular Weight (g/mol) 269.73
MDL Number MFCD00197059
InChI Key ZLGORAQNGFCUFU-UHFFFAOYSA-N
PubChem CID 2769585
IUPAC Name 3-(4-chlorophenyl)-1-phenyl-1H-pyrazol-5-amine
SMILES NC1=CC(=NN1C1=CC=CC=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 192°C
Color Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 269.73
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.