accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSNOONH2

CAS RN 1965305-02-5

IUPAC Name: 2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)-4-nitroaniline
Synonyms: 2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)-4-nitroaniline
Molecular Weight (g/mol): 311.36
Molecular Formula: C16H13N3O2S
InChi Key: UCBKPBJMEXATSY-UHFFFAOYSA-N
SMILES: CC1=C(N=C(S1)C1=C(N)C=CC(=C1)[N+]([O-])=O)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results