accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONNOOCH3H3CCH3ClCl

CAS RN 19666-30-9

IUPAC Name: 5-tert-butyl-3-[2,4-dichloro-5-(propan-2-yloxy)phenyl]-2,3-dihydro-1,3,4-oxadiazol-2-one
Synonyms: oxadiazon
Molecular Weight (g/mol): 345.22
Molecular Formula: C15H18Cl2N2O3
InChi Key: CHNUNORXWHYHNE-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC(N2N=C(OC2=O)C(C)(C)C)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results