accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19690-23-4

IUPAC Name: 6-iodo-7H-purin-2-amine
Synonyms: 6-iodo-7H-purin-2-amine
Molecular Weight (g/mol): 261.03
Molecular Formula: C5H4IN5
InChi Key: CQYPNVKLVHHOSJ-UHFFFAOYSA-N
SMILES: NC1=NC(I)=C2NC=NC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results