accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3CNHOCl

CAS RN 196938-30-4

IUPAC Name: 2-chloro-N-(2,5-dimethoxyphenyl)benzamide
Synonyms: 2-chloro-N-(2,5-dimethoxyphenyl)benzamide
Molecular Weight (g/mol): 291.73
Molecular Formula: C15H14ClNO3
InChi Key: RLHACPNMKPCNMS-UHFFFAOYSA-N
SMILES: COC1=CC(NC(=O)C2=CC=CC=C2Cl)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results