accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 197305-97-8

IUPAC Name: 2-hydrazinyl-4-(trifluoromethyl)pyrimidine
Synonyms: 2-hydrazinyl-4-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 178.12
Molecular Formula: C5H5F3N4
InChi Key: XQCAINVUBSAUBJ-UHFFFAOYSA-N
SMILES: NNC1=NC=CC(=N1)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)