Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydrazino-4-(trifluoromethyl)pyrimidine 98.0+%, TCI America™
SDP

Catalog No. H15745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15745G 5 g
H15741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15745G Supplier TCI America Supplier No. H15745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 197305-97-8
Molecular Formula C5H5F3N4
Molecular Weight (g/mol) 178.118
MDL Number MFCD00115112
InChI Key XQCAINVUBSAUBJ-UHFFFAOYSA-N
Synonym 2-hydrazino-4-trifluoromethyl pyrimidine, 2-hydrazinyl-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ylhydrazine, n-4-trifluoromethyl pyrimidin-2-yl hydrazine, 4-trifluoromethyl pyrimidin-2-yl hydrazine, 1-4-trifluoromethyl pyrimidin-2-yl hydrazine, 2 1h-pyrimidinone, 4-trifluoromethyl-, hydrazone, buttpark 27\08-68, ksc498g6p, 2-hydrazino-4-trifluoromethylpyrimidine
PubChem CID 1268079
IUPAC Name [4-(trifluoromethyl)pyrimidin-2-yl]hydrazine
SMILES C1=CN=C(N=C1C(F)(F)F)NN

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 178.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydrazino-4-(trifluoromethyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.