accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19735-74-1

NH2SN

CAS RN 19735-74-1

IUPAC Name: 4-(adamantan-1-yl)-1,3-thiazol-2-amine
Synonyms: 4-(adamantan-1-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 234.36
Molecular Formula: C13H18N2S
InChi Key: JVJWJUQLRDIVJV-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results