CAS RN 1974-05-6
IUPAC Name:
3-bromoheptane
Synonyms:
3-bromoheptane
Molecular Weight (g/mol):
179.1
Molecular Formula:
C7H15Br
InChi Key:
MLHXKYLLJRLHGH-UHFFFAOYNA-N
SMILES:
CCCCC(Br)CC
1
–
1
of
1
results
1
3-Bromoheptane 93.0+%, TCI America™
CAS: 1974-05-6 Molecular Formula: C7H15Br Molecular Weight (g/mol): 179.10 MDL Number: MFCD00039184 InChI Key: MLHXKYLLJRLHGH-UHFFFAOYNA-N Synonym: heptane, 3-bromo,3-heptyl bromide,acmc-1bscs PubChem CID: 16091 IUPAC Name: 3-bromoheptane SMILES: CCCCC(CC)Br