Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromoheptane 93.0+%, TCI America™
SDP

Catalog No. B123425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B123425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B123425G Supplier TCI America Supplier No. B123425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1974-05-6
Molecular Formula C7H15Br
Molecular Weight (g/mol) 179.10
MDL Number MFCD00039184
InChI Key MLHXKYLLJRLHGH-UHFFFAOYNA-N
Synonym heptane, 3-bromo, 3-heptyl bromide, acmc-1bscs
PubChem CID 16091
IUPAC Name 3-bromoheptane
SMILES CCCCC(CC)Br

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 g
UN Number 1993
Formula Weight 179.10
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromoheptane
EINECSNumber (2)-0067
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.