accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19763-90-7

IUPAC Name: hydrogen (3,4-dichlorophenyl)hydrazine chloride
Synonyms: hydrogen (3,4-dichlorophenyl)hydrazine chloride
Molecular Weight (g/mol): 213.49
Molecular Formula: C6H7Cl3N2
InChi Key: WIKZRYSGQPZTMM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)