Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,4-Dichlorophenyl)hydrazine hydrochloride, 97%, Thermo Scientific™

Catalog No. BTB08664EB
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BTB08664EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB08664EB Supplier Thermo Scientific Chemicals Supplier No. BTB08664EB

missing translation for 'validMainframeMsg'

Specifications

CAS 19763-90-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H7Cl3N2
Linear Formula Cl2C6H3NHNH2·HCl
Quantity 25g
MDL Number MFCD00012937
Synonym 3,4-dichlorophenylhydrazine hydrochloride, 3,4-dichlorophenyl hydrazine hydrochloride, 1-3,4-dichlorophenyl hydrazine hydrochloride, 3,4-dichlorophenyl hydrazine monohydrochloride, hydrazine, 3,4-dichlorophenyl-, monohydrochloride, 3,4-dichlorophenylhydrazinehydrochloride, pubchem7530, acmc-209f1i, ksc491e2j
InChI Key WIKZRYSGQPZTMM-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1NN)Cl)Cl.Cl
IUPAC Name (3,4-dichlorophenyl)hydrazine;hydrochloride
Molecular Weight (g/mol) 213.486
PubChem CID 3084323
Formula Weight 213.49
Percent Purity 97%
Chemical Name or Material 1-(3, 4-Dichlorophenyl)hydrazine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.