accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 197632-75-0

OOOOHOONN(S)CH3H3CCH3H3C

CAS RN 197632-75-0

IUPAC Name: (2S)-3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)propanoic acid
Synonyms: (2S)-3-[1-(tert-butoxycarbonyl)indol-3-yl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}propanoic acid
Molecular Weight (g/mol): 540.62
Molecular Formula: C32H32N2O6
InChi Key: YMZUDCKQUKFBIR-NDEPHWFRSA-N
SMILES: CN([C@@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)C(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)