CAS RN 19764-96-6
IUPAC Name:
1,3,3-trimethyl-2-[(5E)-7-[(2Z)-1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene]hepta-1,3,5-trien-1-yl]-3H-indol-1-ium iodide
Synonyms:
1,3,3-trimethyl-2-[(5E)-7-[(2Z)-1,3,3-trimethylindol-2-ylidene]hepta-1,3,5-trien-1-yl]indol-1-ium iodide
Molecular Weight (g/mol):
536.5
Molecular Formula:
C29H33IN2
InChi Key:
JKXWXYURKUEZHV-UHFFFAOYSA-M
SMILES:
[I-].CN1C2=CC=CC=C2C(C)(C)\C1=C\C=C\C=CC=CC1=[N+](C)C2=CC=CC=C2C1(C)C
1
–
1
of
1
results
1
Sigma Aldrich 1,1',3,3,3',3'-Hexamethylindotricarbocyanine iodide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.