accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOSOOHS

CAS RN 19767-45-4

IUPAC Name: sodium 2-sulfanylethane-1-sulfonate
Synonyms: mesna
Molecular Weight (g/mol): 164.17
Molecular Formula: C2H5NaO3S2
InChi Key: XOGTZOOQQBDUSI-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCS
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (4)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (5)
Grade 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (7)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)