accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19777-66-3

IUPAC Name: propane-1,2-diamine dihydrochloride
Synonyms: propylenediamine dihydrochloride
Molecular Weight (g/mol): 147.04
Molecular Formula: C3H12Cl2N2
InChi Key: AEIAMRMQKCPGJR-UHFFFAOYNA-N
SMILES: Cl.Cl.CC(N)CN
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)