Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1,2-Diaminopropane dihydrochloride, 98%

Catalog No. AAH6075303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6075303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6075303 Supplier Thermo Scientific Chemicals Supplier No. H6075303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 19777-66-3 | C3H12Cl2N2 | 147.043 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19777-66-3
Molecular Formula C3H12Cl2N2
Molecular Weight (g/mol) 147.043
MDL Number MFCD00050706
InChI Key AEIAMRMQKCPGJR-QTNFYWBSSA-N
Synonym s---1,2-diaminopropane dihydrochloride, s-propane-1,2-diamine dihydrochloride, s-1,2-diaminopropane dihydrochloride, s---propylenediamine dihydrochloride, s-1,2-propanediamine dihydrochloride, 2s-propane-1,2-diamine dihydrochloride, s-1,2-diaminopropane 2hcl, s-propylenediamine dihydrochloride, 1,2-propanediamine, dihydrochloride, s, pubchem15232
PubChem CID 12217445
IUPAC Name (2S)-propane-1,2-diamine;dihydrochloride
SMILES CC(CN)N.Cl.Cl

Specifications

Melting Point 227°C to 229°C
Linear Formula CH3CH(NH2)CH2NH2·2HCl
Quantity 1 g
Beilstein 5740936
Optical Rotation −4.0° (c=20 in Water)
Formula Weight 147.05
Percent Purity 99%
Chemical Name or Material (S)-(-)-1,2-Diaminopropane dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.