CAS RN 19781-62-5
IUPAC Name:
4-ethylcyclohexan-1-ol
Synonyms:
4-ethylcyclohexan-1-ol
Molecular Weight (g/mol):
128.22
Molecular Formula:
C8H16O
InChi Key:
RVTKUJWGFBADIN-UHFFFAOYSA-N
SMILES:
CCC1CCC(O)CC1
1
–
1
of
1
results
1
trans-4-Ethylcyclohexanol 96.0+%, TCI America™
CAS: 19781-62-5 Molecular Formula: C8H16O Molecular Weight (g/mol): 128.215 MDL Number: MFCD00070694 InChI Key: RVTKUJWGFBADIN-UHFFFAOYSA-N PubChem CID: 78293 IUPAC Name: 4-ethylcyclohexan-1-ol SMILES: CCC1CCC(CC1)O