Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Ethylcyclohexanol 96.0+%, TCI America™
SDP

Catalog No. E07685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07685G Supplier TCI America Supplier No. E07685G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19781-62-5
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00070694
InChI Key RVTKUJWGFBADIN-UHFFFAOYSA-N
PubChem CID 78293
IUPAC Name 4-ethylcyclohexan-1-ol
SMILES CCC1CCC(CC1)O

Specifications

Color Yellow
Boiling Point 84°C
Quantity 5 g
Formula Weight 128.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-4-Ethylcyclohexanol
EINECSNumber (3)-2301
RTECSNumber GV9050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.