accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(S)NH2OHO

CAS RN 19794-53-7

IUPAC Name: (2S)-2-amino-3-methoxypropanoic acid
Synonyms: (2S)-2-amino-3-methoxypropanoic acid
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: KNTFCRCCPLEUQZ-VKHMYHEASA-N
SMILES: COC[C@H](N)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results