accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClSOONH2

CAS RN 19797-32-1

IUPAC Name: 3,5-dichlorobenzene-1-sulfonamide
Synonyms: 3,5-dichlorobenzenesulfonamide
Molecular Weight (g/mol): 226.07
Molecular Formula: C6H5Cl2NO2S
InChi Key: AHNOVNYOUPQVRX-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC(Cl)=CC(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results