Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorobenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. D35025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35025G Supplier TCI America Supplier No. D35025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19797-32-1
Molecular Formula C6H5Cl2NO2S
Molecular Weight (g/mol) 226.071
MDL Number MFCD00117161
InChI Key AHNOVNYOUPQVRX-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzene-1-sulfonamide, 3,5-dichlorobenzenesulphonamide, benzenesulfonamide, 3,5-dichloro, 3,5-bis chloranyl benzenesulfonamide, pubchem11789, maybridge1_001366, acmc-209f2d, spr_4, benzenesulfonamide,3,5-dichloro, 3,5-dichlorobenzenesulfonamide
PubChem CID 705607
IUPAC Name 3,5-dichlorobenzenesulfonamide
SMILES C1=C(C=C(C=C1Cl)Cl)S(=O)(=O)N

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.07
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorobenzenesulfonamide
RTECSNumber DB1670000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.